Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZUMO3
Entrez Gene ID
100129669
Gene Name
IZUMO family member 3
Gene Aliases
C9orf134, bA20A20.1
Chromosome Location
Chr.9: 24543214 - 24545930 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271706.1 NP_001258635.1 3 - 4 421 86
GenBank mRNA HY236269.1 - 3 - 4 86

View Full Product Details