The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276392.1 | NP_001263321.1 | 1 - 2 | 94 | 114 |
GenBank mRNA | BC127709.1 | - | 1 - 2 | 91 | 114 |