Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 54007298 - 54018186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010978.3 NP_001010978.2 4 - 5 415 78
NM_001276392.1 NP_001263321.1 2 - 3 234 78
NM_001276393.1 NP_001263322.1 3 - 4 286 78
NM_001276395.1 NP_001263324.1 3 - 4 255 78
XM_011541446.1 XP_011539748.1 4 - 5 395 78
XM_017001264.1 XP_016856753.1 3 - 4 238 78
GenBank mRNA BC127708.1 - 4 - 5 412 78
BC127709.1 - 2 - 3 231 78
BC140760.1 - 3 - 4 230 78
BC144466.1 - 3 - 4 230 78
BC171743.1 - 3 - 4 199 78

View Full Product Details