Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 54007298 - 54018186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010978.3 NP_001010978.2 5 - 6 543 102
NM_001276392.1 NP_001263321.1 3 - 4 362 102
NM_001276393.1 NP_001263322.1 4 - 5 414 102
NM_001276394.1 NP_001263323.1 3 - 4 276 102
NM_001276395.1 NP_001263324.1 4 - 5 383 102
XM_011541446.1 XP_011539748.1 5 - 6 523 102
GenBank mRNA BC103740.1 - 3 - 4 264 102
BC127708.1 - 5 - 6 540 102
BC127709.1 - 3 - 4 359 102
BC140760.1 - 4 - 5 358 102
BC144466.1 - 4 - 5 358 102
BC171743.1 - 4 - 5 327 102

View Full Product Details