The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278063.1 | NP_001264992.1 | 4 - 5 | 2809 | 136 |
XM_006722525.3 | XP_006722588.1 | 4 - 5 | 3150 | 136 | |
GenBank mRNA | DW009659.1 | - | 4 - 5 | 293 | 136 |