The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002988.3 | NP_002979.1 | 3 - 3 | 720 | 114 |
GenBank mRNA | AB000221.1 | - | 3 - 3 | 712 | 114 |
BU617136.1 | - | 1 - 1 | 114 | ||
Y13710.1 | - | 3 - 3 | 719 | 114 |