Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NAPB
- Entrez Gene ID
-
63908
- Gene Name
- NSF attachment protein beta
- Gene Aliases
-
SNAP-BETA, SNAPB
- Chromosome Location
-
Chr.20: 23374519 - 23421519 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104266.1 |
|
10 - 11 |
2268 |
67 |
GenBank mRNA |
BX648330.1 |
- |
10 - 11 |
2224 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
GDNF inducible zinc finger protein 1 |
GZF1 |
ZBTB23, ZNF336 |
-1.5kb
|
Anti-sense |
Chr. - 23361585 - 23373062 on Build GRCh38 |
Downstream |
cystatin like 1 |
CSTL1 |
CTES1, RCET11, dJ322G13.4 |
62.2kb
|
Anti-sense |
Chr. - 23439685 - 23444930 on Build GRCh38 |
View Full Product Details