Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MYRIP
- Entrez Gene ID
-
25924
- Gene Name
- myosin VIIA and Rab interacting protein
- Gene Aliases
-
SLAC2-C, SLAC2C
- Chromosome Location
-
Chr.3: 39808914 - 40260321 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104316.1 |
|
11 - 12 |
2198 |
69 |
GenBank mRNA |
BC109312.1 |
- |
11 - 12 |
2026 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
EIF1B antisense RNA 1 |
EIF1B-AS1 |
|
|
Anti-sense |
Chr. - 40173145 - 40309698 on Build GRCh38 |
Downstream |
eukaryotic translation initiation factor 1B |
EIF1B |
GC20 |
75.6kb
|
Sense |
Chr. - 40309682 - 40312424 on Build GRCh38 |
View Full Product Details