Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC25A44
- Entrez Gene ID
-
9673
- Gene Name
- solute carrier family 25 member 44
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 156193711 - 156212796 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104412.1 |
|
1 - 2 |
322 |
76 |
GenBank mRNA |
AK309690.1 |
- |
1 - 2 |
151 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA911335.1 |
- |
1 - 2 |
151 |
|
Chr. 1 - 156194103 - 156210674 [+] on Build GRCh38 |
76 |
GenBankmRNA |
DA716953.1 |
- |
1 - 2 |
151 |
|
Chr. 1 - 156194103 - 156210647 [+] on Build GRCh38 |
76 |
GenBankmRNA |
DA590482.1 |
- |
1 - 2 |
137 |
|
Chr. 1 - 156194117 - 156210689 [+] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
semaphorin 4A |
SEMA4A |
CORD10, RP35, SEMAB, SEMB |
-16.2kb
|
Sense |
Chr. - 156149673 - 156177751 on Build GRCh38 |
Downstream |
polyamine-modulated factor 1 |
PMF1 |
|
192.0
|
Sense |
Chr. - 156212988 - 156240077 on Build GRCh38 |
View Full Product Details