Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC648987
- Entrez Gene ID
-
648987
- Gene Name
- uncharacterized LOC648987
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 43014729 - 43067439 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104649.1 |
|
1 - 2 |
1734 |
115 |
GenBank mRNA |
BC039088.1 |
- |
1 - 2 |
1734 |
115 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC643977 |
FLJ32255 |
|
-23.0kb
|
Sense |
Chr. - 42985398 - 42993333 on Build GRCh38 |
Downstream |
annexin A2 receptor |
ANXA2R |
AX2R, AXIIR, C5orf39 |
20.3kb
|
Sense |
Chr. - 43039080 - 43040345 on Build GRCh38 |
View Full Product Details