Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PAIP2
- Entrez Gene ID
-
51247
- Gene Name
- poly(A) binding protein interacting protein 2
- Gene Aliases
-
PAIP-2, PAIP2A
- Chromosome Location
-
Chr.5: 139341832 - 139369720 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109830.1 |
|
2 - 3 |
310 |
124 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD556896.1 |
- |
2 - 3 |
241 |
|
Chr. 5 - 139341901 - 139369102 [+] on Build GRCh38 |
124 |
GenBankmRNA |
CD517972.1 |
- |
|
319 |
|
Chr. - - [-] on Build |
124 |
GenBankmRNA |
BM701858.1 |
- |
2 - 3 |
277 |
|
Chr. 5 - 139341884 - 139368910 [+] on Build GRCh38 |
124 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
matrin 3 |
MATR3 |
ALS21, MPD2, VCPDM |
-32.1kb
|
Sense |
Chr. - 139273752 - 139331677 on Build GRCh38 |
Downstream |
solute carrier family 23 member 1 |
SLC23A1 |
SLC23A2, SVCT1, YSPL3 |
2.5kb
|
Anti-sense |
Chr. - 139367196 - 139384553 on Build GRCh38 |
View Full Product Details