Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ST7-AS2
- Entrez Gene ID
-
93654
- Gene Name
- ST7 antisense RNA 2
- Gene Aliases
-
ST7AS2, ST7OT2
- Chromosome Location
-
Chr.7: 117072072 - 117145592 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109981.1 |
|
2 - 3 |
276 |
78 |
GenBank mRNA |
DB054726.1 |
- |
2 - 3 |
276 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 6132 |
MIR6132 |
hsa-mir-6132 |
-98.0kb
|
Anti-sense |
Chr. - 117020211 - 117020319 on Build GRCh38 |
Downstream |
ST7 overlapping transcript 3 |
ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
38.4kb
|
Anti-sense |
Chr. - 117182681 - 117209937 on Build GRCh38 |
View Full Product Details