Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZFAT
- Entrez Gene ID
-
57623
- Gene Name
- zinc finger and AT-hook domain containing
- Gene Aliases
-
AITD3, ZFAT1, ZNF406
- Chromosome Location
-
Chr.8: 134477788 - 134713049 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110323.1 |
|
12 - 13 |
3334 |
95 |
GenBank mRNA |
AL832066.1 |
- |
12 - 13 |
3334 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE747464.1 |
- |
3 - 4 |
413 |
|
Chr. 8 - 134512571 - 134583949 [-] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927822 |
LOC101927822 |
|
-630.4kb
|
Anti-sense |
Chr. - 133886496 - 133902407 on Build GRCh38 |
Downstream |
ZFAT antisense RNA 1 |
ZFAT-AS1 |
NCRNA00070, SAS-ZFAT, ZFAT-AS, ZFATAS |
32.9kb
|
Anti-sense |
Chr. - 134598071 - 134600689 on Build GRCh38 |
View Full Product Details