Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PCSK9
- Entrez Gene ID
-
255738
- Gene Name
- proprotein convertase subtilisin/kexin type 9
- Gene Aliases
-
FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9
- Chromosome Location
-
Chr.1: 55039476 - 55064853 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110451.1 |
|
6 - 7 |
962 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
barttin CLCNK type accessory beta subunit |
BSND |
BART, DFNB73 |
-49.2kb
|
Sense |
Chr. - 54998944 - 55008792 on Build GRCh38 |
Downstream |
ubiquitin specific peptidase 24 |
USP24 |
|
7.7kb
|
Anti-sense |
Chr. - 55066359 - 55215374 on Build GRCh38 |
View Full Product Details