Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LGR5
- Entrez Gene ID
-
8549
- Gene Name
- leucine rich repeat containing G protein-coupled receptor 5
- Gene Aliases
-
FEX, GPR49, GPR67, GRP49, HG38
- Chromosome Location
-
Chr.12: 71439770 - 71586310 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110596.1 |
|
19 - 20 |
2493 |
78 |
GenBank mRNA |
AK075399.1 |
- |
19 - 20 |
2279 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tetraspanin 8 |
TSPAN8 |
CO-029, TM4SF3 |
-425.6kb
|
Anti-sense |
Chr. - 71125093 - 71157999 on Build GRCh38 |
Downstream |
zinc finger C3H1-type containing |
ZFC3H1 |
CCDC131, CSRC2, PSRC2 |
23.3kb
|
Anti-sense |
Chr. - 71609599 - 71663969 on Build GRCh38 |
View Full Product Details