Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01612
- Entrez Gene ID
-
101928223
- Gene Name
- long intergenic non-protein coding RNA 1612
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 170226591 - 170283723 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125889.1 |
|
1 - 2 |
196 |
119 |
View Full Product Details