Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NTM-IT
- Entrez Gene ID
-
100874281
- Gene Name
- NTM intronic transcript
- Gene Aliases
-
NTM-IT3
- Chromosome Location
-
Chr.11: 132284302 - 132288459 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046820.1 |
|
1 - 2 |
60 |
61 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NTM antisense RNA 1 |
NTM-AS1 |
C11orf39 |
-620.7kb
|
Anti-sense |
Chr. - 131662129 - 131663556 on Build GRCh38 |
Downstream |
opioid binding protein/cell adhesion molecule like |
OPCML |
IGLON1, OBCAM, OPCM |
129.9kb
|
Anti-sense |
Chr. - 132403361 - 133532983 on Build GRCh38 |
View Full Product Details