Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA5
Entrez Gene ID
4698
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Gene Aliases
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13
Chromosome Location
Chr.7: 123536997 - 123601651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104168.2 2 - 3 175 84
GenBank mRNA BG400546.1 - 2 - 3 76 84

View Full Product Details