Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMS1P5
Entrez Gene ID
399761
Gene Name
BMS1, ribosome biogenesis factor pseudogene 5
Gene Aliases
BMS1LP1, BMS1LP5, BMS1P1, bA556L1.3
Chromosome Location
Chr.10: 46786733 - 46811989 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003611.2 8 - 9 1237 101
GenBank mRNA AK307333.1 - 8 - 9 1096 101
BC017177.2 - 7 - 8 1085 101


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ948157.1 - 2 - 3 214 Chr. 10 - 46796716 - 46835830 [+] on Build GRCh38 101
GenBankmRNA BU527262.1 - 2 - 3 215 Chr. 10 - 46796716 - 46836378 [+] on Build GRCh38 101
GenBankmRNA BQ933440.1 - 214 Chr. - - [-] on Build 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream glutamate dehydrogenase 1 pseudogene 7 GLUD1P7 GLUD1P2, GLUDP2, GLUDP7, bA342C24.3 -11.3kb Anti-sense Chr. - 46759023 - 46786770 on Build GRCh38
Downstream FERM and PDZ domain containing 2B, pseudogene FRMPD2B FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2 64.7kb Sense Chr. - 46876674 - 46895326 on Build GRCh38

View Full Product Details