Product Details

Assay Reference Genome
Location

Chr.4:89730533 on build GRCh38
Cytoband
4q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6622
Gene Symbol
SNCA
Gene Name
synuclein alpha
Gene Aliases
NACP, PARK1, PARK4, PD1
Location
Chr.4:89724099-89838324 on build GRCh38
Assay Gene Location
Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SNCA NM_000345.3 NP_000336.1
NM_001146054.1 NP_001139526.1
NM_001146055.1 NP_001139527.1
NM_007308.2 NP_009292.1
XM_011532208.2 XP_011530510.1
XM_017008562.1 XP_016864051.1
XM_017008563.1 XP_016864052.1
AK290169.1
AY049786.1 AAL15443.1
BC013293.2 AAH13293.1
BC108275.1 AAI08276.1
CR457058.1 CAG33339.1
CR541653.1 CAG46454.1
D31839.1 BAA06625.1
DA391115.1
JN709859.1
JN709860.1
JN709861.1
JN709862.1
JN709863.1
JN709864.1
JN709865.1
JN709866.1
JN709867.1
JN709868.1
JN709869.1
L08850.1 AAA16117.1
L36674.1 AAA98493.1
L36675.1 AAA98487.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1012406 Chr4:89246630 - 90245636 on Build GRCh38 Loss GPRIN3 SNCA CCSER1 SNCA-AS1 MMRN1
nsv594806 Chr4:88621130 - 90539537 on Build GRCh38 Gain GPRIN3 FAM13A SNCA CCSER1 SNCA-AS1 MMRN1 TIGD2 FAM13A-AS1 HERC3 NAP1L5

View Full Product Details