Product Details

Assay Reference Genome
Location

Chr.11:113423758 on build GRCh38
Cytoband
11q23.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1813
Gene Symbol
DRD2
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
Location
Chr.11:113409595-113475279 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DRD2 NM_000795.3 NP_000786.1
NM_016574.3 NP_057658.2
XM_017017296.1 XP_016872785.1
AB209832.1 BAD93069.1
AB451480.1
AF176812.1 AAF61479.1
AK308777.1
BC021195.2 AAH21195.1
BF437994.1
M29066.1 AAA52761.1
M30625.1 AAA88024.1
S62137.1 AAB26819.1
S69899.1 AAB20571.1
X51362.1 CAA35746.1
X51645.1 CAB56463.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv556444 Chr11:113318201 - 113512672 on Build GRCh38 Gain TTC12 ANKK1 MIR4301 DRD2

View Full Product Details