Product Details

Assay Reference Genome
Location

Chr.15:58445283 on build GRCh38
Cytoband
15q21.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3990
Gene Symbol
LIPC
Gene Name
lipase C, hepatic type
Gene Aliases
HDLCQ12, HL, HTGL, LIPH
Location
Chr.15:58410754-58568952 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LIPC NM_000236.2 NP_000227.2
XM_005254372.1 XP_005254429.1
AK290062.1
AK292631.1
AK293324.1
AK309448.1
AK315306.1
BC132825.1
BC136495.1
BC146659.1
D83548.1 BAA12014.1
J03540.1 AAA59520.1
J03895.1 AAA61165.1
X07228.1 CAA30188.1
Entrez Gene ID
101928694
Gene Symbol
LIPC-AS1
Gene Name
LIPC antisense RNA 1
Gene Aliases
Location
Chr.15:58434901-58498735 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LIPC-AS1 NR_120338.1
AK091917.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv569600 Chr15:58439368 - 58486840 on Build GRCh38 Loss LIPC LIPC-AS1
nsv569598 Chr15:58260407 - 58560264 on Build GRCh38 Loss LOC101928668 LIPC LIPC-AS1 LOC101928635
nsv1043362 Chr15:58434138 - 58464372 on Build GRCh38 Loss LIPC LIPC-AS1
esv3581729 Chr15:58432325 - 58464372 on Build GRCh38 Loss LIPC LIPC-AS1
nsv1047048 Chr15:58409201 - 58492601 on Build GRCh38 Loss LIPC LIPC-AS1 LOC101928635

View Full Product Details