Product Details

Assay Reference Genome
Location

Chr.16:23857365 on build GRCh38
Cytoband
16p12.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5579
Gene Symbol
PRKCB
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
Location
Chr.16:23835979-24220611 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PRKCB NM_002738.6 NP_002729.2
NM_212535.2 NP_997700.1
BC036472.1 AAH36472.1
BI549165.1
DB456157.1
M13975.1 AAA60095.1
X06318.1 CAA29634.1
X07109.1 CAA30130.1

View Full Product Details