Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
UVSSA | NM_001317934.1 | NP_001304863.1 | ||
NM_001317935.1 | NP_001304864.1 | |||
NM_020894.3 | NP_065945.2 | |||
XM_017008490.1 | XP_016863979.1 | |||
XM_017008491.1 | XP_016863980.1 | |||
XM_017008492.1 | XP_016863981.1 | |||
XM_017008493.1 | XP_016863982.1 | |||
XM_017008494.1 | XP_016863983.1 | |||
XM_017008495.1 | XP_016863984.1 | |||
XM_017008496.1 | 9 | 4141 | XP_016863985.1 | |
XM_017008498.1 | XP_016863987.1 | |||
AB040963.1 | ||||
BC110331.1 | AAI10332.1 | |||
BC140901.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr4:546316 - 1420011 on Build GRCh38 | Loss | TMED11P TMEM175 LOC107986211 LOC101928521 MYL5 LOC101928548 MFSD7 SPON2 SLC26A1 ATP5I CTBP1 DGKQ CTBP1-AS2 LOC105374344 RNF212 PCGF3 UVSSA NKX1-1 CPLX1 GAK MAEA FGFRL1 LOC100130872 CTBP1-AS IDUA LOC100129917 CRIPAK PDE6B |
nsv593278 | Chr4:1165892 - 1421910 on Build GRCh38 | Loss | UVSSA LOC100130872 NKX1-1 CTBP1 LOC101928548 CTBP1-AS CTBP1-AS2 CRIPAK SPON2 MAEA |
nsv593301 | Chr4:1306289 - 1358897 on Build GRCh38 | Loss | UVSSA MAEA |
nsv593302 | Chr4:1344830 - 1420896 on Build GRCh38 | Loss | UVSSA NKX1-1 CRIPAK |
nsv508988 | Chr4:1190955 - 1358870 on Build GRCh38 | Insertion | UVSSA LOC100130872 CTBP1 LOC101928548 CTBP1-AS CTBP1-AS2 SPON2 MAEA |
nsv593281 | Chr4:1209477 - 1358897 on Build GRCh38 | Loss | UVSSA CTBP1 CTBP1-AS CTBP1-AS2 MAEA |