Product Details

Assay Reference Genome
Location

Chr.5:78023556 on build GRCh38
Cytoband
5q14.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8546
Gene Symbol
AP3B1
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
Location
Chr.5:78002326-78294755 on build GRCh38
Assay Gene Location
Within Intron 25
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
AP3B1 NM_001271769.1 NP_001258698.1
NM_003664.4 NP_003655.3
XM_005248618.3 XP_005248675.1
XM_017010001.1 XP_016865490.1
AK225908.1
AK291201.1
BC038444.1 AAH38444.1
BX538041.1 CAD97982.1
KU178426.1
KU178427.1
KU178428.1
U81504.1 AAB61638.1
U91931.1 AAD03778.1

View Full Product Details