Product Details

Assay Reference Genome
Location

Chr.11:95844204 on build GRCh38
Cytoband
11q21
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8898
Gene Symbol
MTMR2
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
Location
Chr.11:95832880-95924207 on build GRCh38
Assay Gene Location
Within Intron 15
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MTMR2 NM_001243571.1 NP_001230500.1
NM_016156.5 NP_057240.3
NM_201278.2 NP_958435.1
NM_201281.2 NP_958438.1
XM_005274374.2 XP_005274431.1
XM_005274375.2 XP_005274432.1
XM_006718934.2 XP_006718997.1
XM_006718935.2 XP_006718998.1
XM_006718936.3 XP_006718999.1
XM_011543058.2 XP_011541360.1
XM_011543059.2 XP_011541361.1
XM_017018517.1 XP_016874006.1
XM_017018518.1 XP_016874007.1
AB028996.1
AK027038.1
AK291277.1
AK302940.1
BC052990.1 AAH52990.1
BX647997.1
U58033.1 AAC79118.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv832235 Chr11:95753612 - 95935071 on Build GRCh38 Gain FAM76B CEP57 MTMR2

View Full Product Details