Product Details

Assay Reference Genome
Location

Chr.14:21293300 on build GRCh38
Cytoband
14q11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
57096
Gene Symbol
RPGRIP1
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
Location
Chr.14:21287977-21351316 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
RPGRIP1 NM_020366.3 NP_065099.3
AJ417067.1 CAD01135.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv509527 Chr14:21258526 - 21302138 on Build GRCh38 Insertion RPGRIP1 HNRNPC

View Full Product Details