The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000201.2 | NP_000192.2 | 5 - 6 | 1502 | 99 |
GenBank mRNA | AF340038.1 | - | 5 - 6 | 1183 | 99 |
AF340039.1 | - | 5 - 6 | 1171 | 99 | |
AK130659.1 | - | 5 - 6 | 1223 | 99 | |
AK298054.1 | - | 5 - 6 | 1109 | 99 | |
AK298983.1 | - | 5 - 6 | 935 | 99 | |
AK301412.1 | - | 3 - 4 | 584 | 99 | |
AK312636.1 | - | 5 - 6 | 1223 | 99 | |
BC015969.2 | - | 5 - 6 | 1254 | 99 | |
BT006854.1 | - | 5 - 6 | 1183 | 99 | |
J03132.1 | - | 5 - 6 | 1240 | 99 | |
M24283.1 | - | 5 - 6 | 1254 | 99 | |
M55038.1 | - | 2 - 3 | 415 | 99 | |
X06990.1 | - | 5 - 6 | 1195 | 99 |