Gene Transcripts

Gene Symbol
Slc9a3
Entrez Gene ID
105243
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 3
Gene Aliases
9030624O13Rik, AI930210, NHE-3, NHE3
Chromosome Location
Chr. 13: 74086254 - 74169442 on Build GRCm38
UniGene ID
Mm.261564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081060.1 NP_001074529.1 16 2353
GenBank mRNA AK033564.1 - 4 455

View Full Product Details