Gene Transcripts

Gene Symbol
Cela1
Entrez Gene ID
109901
Gene Name
chymotrypsin-like elastase family, member 1
Gene Aliases
1810009A17Rik, 1810062B19Rik, Ela-1, Ela1, PC-TsF
Chromosome Location
Chr. 15: 100674422 - 100687920 on Build GRCm38
UniGene ID
Mm.2131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033612.2 NP_291090.2 4 320
GenBank mRNA AK007392.1 - 4 318
BC011218.1 - 4 294 4208442
CB951068.1 - 4 320
M27347.1 - 3 203

View Full Product Details