Gene Transcripts
- Gene Symbol
- Ppp2r4
- Entrez Gene ID
- 110854
- Gene Name
- protein phosphatase 2A activator, regulatory subunit B
- Gene Aliases
- 2610042B21Rik, C77440, N28142, PR53
- Chromosome Location
- Chr. 2: 30416050 - 30447807 on Build GRCm38
- UniGene ID
- Mm.275393
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138748.5 |
NP_620087.3
|
8, 9 |
1029 |
|
GenBank mRNA |
AK031591.1 |
- |
8, 9 |
1030 |
|
AK046288.1 |
- |
8, 9 |
833 |
|
AK049184.1 |
- |
8, 9 |
1020 |
|
AK150029.1 |
- |
8, 9 |
1024 |
|
AK160980.1 |
- |
7, 8 |
2194 |
|
AK205670.1 |
- |
1, 2 |
50 |
|
AK206672.1 |
- |
1, 2 |
50 |
|
AK217237.1 |
- |
1, 2 |
50 |
|
AK218137.1 |
- |
|
50 |
|
AY035997.1 |
- |
8, 9 |
882 |
|
BC006626.1 |
- |
8, 9 |
945 |
3158086 |
BC052852.1 |
- |
8, 9 |
955 |
30058414 |
- Gene Symbol
- Qars
- Entrez Gene ID
- 97541
- Gene Name
- glutaminyl-tRNA synthetase
- Gene Aliases
- 1110018N24Rik, 1200016L19Rik, C80286, GLNRS
- Chromosome Location
- Chr. 9: 108508005 - 108515941 on Build GRCm38
- UniGene ID
- Mm.272427
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK189159.1 |
- |
|
277 |
|
View Full Product Details