Gene Transcripts

Gene Symbol
Chrm4
Entrez Gene ID
12672
Gene Name
cholinergic receptor, muscarinic 4
Gene Aliases
Chrm-4, M4
Chromosome Location
Chr. 2: 91919619 - 91929835 on Build GRCm38
UniGene ID
Mm.330405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007699.2 NP_031725.1 2 1381
GenBank mRNA BC120504.1 - 1 1225
BC137677.1 - 1 1225 8860696

View Full Product Details