Gene Transcripts

Gene Symbol
Enam
Entrez Gene ID
13801
Gene Name
enamelin
Gene Aliases
abte
Chromosome Location
Chr. 5: 88487975 - 88506049 on Build GRCm38
UniGene ID
Mm.8014
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017468.3 NP_059496.1 9 869
GenBank mRNA BC115848.1 - 9 781
U82698.1 - 9 870

View Full Product Details