Gene Transcripts

Gene Symbol
F9
Entrez Gene ID
14071
Gene Name
coagulation factor IX
Gene Aliases
AW111646, Cf-9, Cf9
Chromosome Location
Chr. X: 59999464 - 60030758 on Build GRCm38
UniGene ID
Mm.391283
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK149372.1 - 6 546
BC125617.1 - 6 544
BC132393.1 - 6 544
M23109.1 - 6 509

View Full Product Details