Gene Transcripts

Gene Symbol
Gpr6
Entrez Gene ID
140741
Gene Name
G protein-coupled receptor 6
Gene Aliases
AI852874, Gm233
Chromosome Location
Chr. 10: 41069974 - 41072698 on Build GRCm38
UniGene ID
Mm.290693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199058.1 NP_951013.1 1 831
GenBank mRNA AK139367.1 - 2 1010
AY064488.1 - 1 831
BC111861.1 - 1 820

View Full Product Details