Gene Transcripts

Gene Symbol
Fabp2
Entrez Gene ID
14079
Gene Name
fatty acid binding protein 2, intestinal
Gene Aliases
Fabpi, I-FABP
Chromosome Location
Chr. 3: 122895072 - 122899506 on Build GRCm38
UniGene ID
Mm.28398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007980.3 NP_032006.1 2 468
GenBank mRNA AA245078.1 - 2 468
AK007995.1 - 2 166
AY523818.1 - 2 154
AY523819.1 - 2 154
BC013457.1 - 2 148 4218050
BY707992.1 - 2 164
M65034.1 - 2 162

View Full Product Details