Gene Transcripts

Gene Symbol
Fpr-rs4
Entrez Gene ID
14291
Gene Name
formyl peptide receptor, related sequence 4
Gene Aliases
Chromosome Location
Chr. 17: 18021733 - 18022704 on Build GRCm38
UniGene ID
Mm.455331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008041.2 NP_032067.2 1 176
GenBank mRNA BC115534.1 - 1 172
BC115535.1 - 1 176

View Full Product Details