Gene Transcripts
- Gene Symbol
- Hipk1
- Entrez Gene ID
- 15257
- Gene Name
- homeodomain interacting protein kinase 1
- Gene Aliases
- 1110062K04Rik, Myak
- Chromosome Location
- Chr. 3: 103739815 - 103791894 on Build GRCm38
- UniGene ID
- Mm.20827
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301304.1 |
NP_001288233.1
|
7, 8 |
1923 |
|
NM_001301306.1 |
NP_001288235.1
|
7, 8 |
2033 |
|
NM_010432.2 |
NP_034562.2
|
7, 8 |
2033 |
|
GenBank mRNA |
AF071070.1 |
- |
7, 8 |
1923 |
|
AF071071.1 |
- |
7, 8 |
1921 |
|
AF077658.1 |
- |
7, 8 |
1937 |
|
AF170303.1 |
- |
7, 8 |
1833 |
|
AF170304.1 |
- |
7, 8 |
1833 |
|
AK134408.1 |
- |
7, 8 |
2034 |
|
BC145697.1 |
- |
7, 8 |
1786 |
40131000 |
BC145699.1 |
- |
7, 8 |
1786 |
40131002 |
DQ279086.1 |
- |
1, 2 |
117 |
|
DQ279087.1 |
- |
1, 2 |
119 |
|
View Full Product Details