Gene Transcripts

Gene Symbol
Lama5
Entrez Gene ID
16776
Gene Name
laminin, alpha 5
Gene Aliases
AA408760, AA408762, AI853660, [a]5, laminin-511, mKIAA0533
Chromosome Location
Chr. 2: 180176373 - 180225905 on Build GRCm38
UniGene ID
Mm.4339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081171.2 NP_001074640.1 60 8197
GenBank mRNA AK129157.1 - 21 2971
CK021961.1 - 6 649
U37501.1 - 60 7891

View Full Product Details