Gene Transcripts

Gene Symbol
Pfkm
Entrez Gene ID
18642
Gene Name
phosphofructokinase, muscle
Gene Aliases
AI131669, ATP-PFK, PFK-A, PFK-M, Pfk-4, Pfk4, Pfka, Pfkx
Chromosome Location
Chr. 15: 98092589 - 98132447 on Build GRCm38
UniGene ID
Mm.272582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163487.1 NP_001156959.1 21 2186
NM_001163488.1 NP_001156960.1 21 2122
NM_021514.4 NP_067489.3 20 2139
GenBank mRNA AF249894.1 - 20 2079
AK002711.2 - 21 2089
AK049773.1 - 21 2094
BC005526.1 - 20 2103 3495173

View Full Product Details