Gene Transcripts

Gene Symbol
Dcdc2a
Entrez Gene ID
195208
Gene Name
doublecortin domain containing 2a
Gene Aliases
AW492955, Dcdc2, RU2
Chromosome Location
Chr. 13: 25020435 - 25210706 on Build GRCm38
UniGene ID
Mm.284719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195617.1 NP_001182546.1 5 872
NM_177577.3 NP_808245.2 5 872
GenBank mRNA AK220370.1 - 5 645
BC045136.1 - 5 872 6306251

View Full Product Details