Gene Transcripts

Gene Symbol
Fbxo40
Entrez Gene ID
207215
Gene Name
F-box protein 40
Gene Aliases
9830003A13Rik, mKIAA1195
Chromosome Location
Chr. 16: 36963461 - 36990535 on Build GRCm38
UniGene ID
Mm.211899
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037321.1 NP_001032398.1 1 33
GenBank mRNA BC107406.1 - 1 33

View Full Product Details