Gene Transcripts

Gene Symbol
Wdr11
Entrez Gene ID
207425
Gene Name
WD repeat domain 11
Gene Aliases
2900055P10Rik, AW489876, Brwd2, mKIAA1351
Chromosome Location
Chr. 7: 129591714 - 129635738 on Build GRCm38
UniGene ID
Mm.229323
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172255.3 NP_758459.2 5 782
GenBank mRNA AK043051.1 - 3 400
AK166369.1 - 5 738
AK170058.1 - 5 730
BC037177.1 - 5 719 4948388

View Full Product Details