Gene Transcripts

Gene Symbol
Myo5c
Entrez Gene ID
208943
Gene Name
myosin VC
Gene Aliases
9130003O20Rik, BB085560, C330029I24
Chromosome Location
Chr. 9: 75231725 - 75306263 on Build GRCm38
UniGene ID
Mm.41339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081322.1 NP_001074791.1 36, 37 4523
GenBank mRNA AK033571.1 - 12, 13 1328
BC003985.1 - 7, 8 979

View Full Product Details