Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011692.2 | NP_035822.2 | 5 | 536 | |
GenBank mRNA | AK002282.1 | - | 5 | 506 | |
AK008847.1 | - | 5 | 506 | ||
AK077923.1 | - | 5 | 521 | ||
AK151527.1 | - | 5 | 506 | ||
AK153069.1 | - | 5 | 505 | ||
AK159284.1 | - | 5 | 506 | ||
AK167064.1 | - | 5 | 506 | ||
AK169185.1 | - | 5 | 506 | ||
BC132169.1 | - | 5 | 479 | ||
BC132171.1 | - | 5 | 479 | ||
DV066998.1 | - | 5 | 569 | ||
U96760.1 | - | 5 | 487 |