Gene Transcripts
- Gene Symbol
- Dars
- Entrez Gene ID
- 226414
- Gene Name
- aspartyl-tRNA synthetase
- Gene Aliases
- 5730439G15Rik
- Chromosome Location
- Chr. 1: 128363707 - 128417416 on Build GRCm38
- UniGene ID
- Mm.28693
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145507.2 |
NP_663482.2
|
1, 2 |
263 |
|
NM_177445.5 |
NP_803228.2
|
1, 2 |
263 |
|
GenBank mRNA |
AK077533.1 |
- |
1, 2 |
186 |
|
AK077820.1 |
- |
1, 2 |
217 |
|
AK144470.1 |
- |
1, 2 |
208 |
|
AK146350.1 |
- |
1, 2 |
207 |
|
AK151027.1 |
- |
1, 2 |
207 |
|
AK151897.1 |
- |
1, 2 |
174 |
|
AK152304.1 |
- |
1, 2 |
174 |
|
AK152562.1 |
- |
1, 2 |
174 |
|
AK153256.1 |
- |
1, 2 |
174 |
|
AK153299.1 |
- |
1, 2 |
174 |
|
AK166009.1 |
- |
1, 2 |
174 |
|
AK169247.1 |
- |
1, 2 |
174 |
|
AK169716.1 |
- |
1, 2 |
151 |
|
BB576692.2 |
- |
1, 2 |
217 |
|
BC008638.1 |
- |
1, 2 |
161 |
3586278 |
BY276340.1 |
- |
1, 2 |
264 |
|
BY308833.1 |
- |
1, 2 |
261 |
|
View Full Product Details