Gene Transcripts
- Gene Symbol
- Mapkap1
- Entrez Gene ID
- 227743
- Gene Name
- mitogen-activated protein kinase associated protein 1
- Gene Aliases
- AI591529, D230039K05Rik, Sin1
- Chromosome Location
- Chr. 2: 34406771 - 34624958 on Build GRCm38
- UniGene ID
- Mm.270866
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290625.1 |
NP_001277554.1
|
11, 12 |
1581 |
|
NM_001290626.1 |
NP_001277555.1
|
10, 11 |
1579 |
|
NM_177345.4 |
NP_796319.1
|
11, 12 |
1907 |
|
GenBank mRNA |
AK027932.1 |
- |
6, 7 |
680 |
|
AK042154.1 |
- |
10, 11 |
1569 |
|
AK052045.1 |
- |
11, 12 |
1575 |
|
AK132263.1 |
- |
11, 12 |
1571 |
|
BC027377.1 |
- |
6, 7 |
709 |
|
BC031579.1 |
- |
6, 7 |
709 |
|
BC043296.1 |
- |
11, 12 |
1806 |
|
BC090644.1 |
- |
10, 11 |
1481 |
30629263 |
BC096618.1 |
- |
11, 12 |
1880 |
6392317 |
View Full Product Details