Gene Transcripts

Gene Symbol
Tmem69
Entrez Gene ID
230657
Gene Name
transmembrane protein 69
Gene Aliases
A630048M13Rik
Chromosome Location
Chr. 4: 116551528 - 116557042 on Build GRCm38
UniGene ID
Mm.291484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177670.4 NP_808338.2 3 713
GenBank mRNA AK041960.1 - 3 698
BC106178.1 - 3 706 6512571

View Full Product Details