Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153526.5 | NP_705746.1 | 3 | 639 | |
GenBank mRNA | AF527630.1 | - | 3 | 653 | |
AK088380.1 | - | 3 | 686 | ||
AK135941.1 | - | 3 | 690 | ||
AK135989.1 | - | 3 | 692 | ||
AK139913.1 | - | 3 | 689 | ||
AK151186.1 | - | 3 | 689 | ||
BC079559.1 | - | 3 | 639 | ||
BC132167.1 | - | 3 | 647 | 40130444 | |
BC132499.1 | - | 3 | 647 | 40130776 |