Gene Transcripts

Gene Symbol
Sugp2
Entrez Gene ID
234373
Gene Name
SURP and G patch domain containing 2
Gene Aliases
Sfrs14, Srsf14, mKIAA0365
Chromosome Location
Chr. 8: 70234209 - 70263106 on Build GRCm38
UniGene ID
Mm.284505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168290.1 NP_001161762.1 3 1269
NM_172755.3 NP_766343.3 3 1269
GenBank mRNA AF518875.1 - 3 1262
AK042293.1 - 3 1270
AK137294.1 - 3 873
BC057305.1 - 3 923 6822887

View Full Product Details